
2-Bromo-m-xylene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:576-22-7
:
Formula:C₈H₉Br
MDL Number:MFCD00000075
:185.06 g/mol
Boiling Pt:206 °C (1013 hPa)
Melting Pt:–10 °C
Density:1.38 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:209-397-7
:GHS07
Storage Temperature:Ambient