Order Entry
Export
Orders LinkContactUsLinkComponent
α-Bromo-p-xylene, Sigma-Aldrich®

α-Bromo-p-xylene, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIALB83606-25G
Corrosive
Supplier:   Merck
α-Bromo-p-xylene, Sigma-Aldrich®
Catalog # SIALB83606-25G
Supplier:  Merck
CAS Number:  104-81-4
α-Bromo-p-xylene, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  104-81-4
Price
67,20 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  alpha-Bromo-p-xylene, 4-Methylbenzyl bromide, 4-(Bromomethyl)toluene
  • CAS Number:104-81-4
  • Formula:C₈H₉Br
  • MDL Number:MFCD00000181
  • Molecular Weight:185.06 g/mol
  • Merck Index:12,10223
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:38 °C
  • Density:1.324 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:203-240-6
  • UN:3417
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  •  :Density: 1.324 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications