
α-Bromo-o-xylene, Sigma-Aldrich®
Catalog # SIALB83401-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:185.06 g/mol
Merck Index:12,10223
Boiling Pt:216 °C (1013 hPa)
Melting Pt:18…20 °C
Density:1.381 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-951-6
UN:1701
ADR:6.1,II
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:89-92-9
Formula:C₈H₉Br
MDL Number:MFCD00000174