2,5-Dibromo-p-xylene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1074-24-4
:
Formula:C₈H₈Br₂
MDL Number:MFCD00000091
:263.96 g/mol
Boiling Pt:261 °C (1013 hPa)
Melting Pt:75 °C
EINECS:214-038-2
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100