
α,α'-Dibromo-m-xylene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:626-15-3
:
Formula:C₈H₈Br₂
MDL Number:MFCD00000178
:263.96 g/mol
Boiling Pt:135…140 °C (20 mmHg)
Melting Pt:74…76 °C
Density:1.959 g/cm³ (0 °C)
Storage Temperature:Ambient
EINECS:210-931-6
UN:3261
ADR:8,II
:GHS05