1,2-Diphenylethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:103-29-7
:
Formula:(C₆H₅)CH₂CH₂(C₆H₅)
MDL Number:MFCD00004796
:182.27 g/mol
Merck Index:12,01245
Boiling Pt:284 °C (1013 hPa)
Melting Pt:52…54 °C
Density:1.12 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:203-096-4