1,2-Diphenylethane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103-29-7
:
Formula:(C₆H₅)CH₂CH₂(C₆H₅)
MDL Number:MFCD00004796
:182.27 g/mol
Merck Index:14,1198
Boiling Pt:284 °C
Melting Pt:50…53 °C
Density:1.013
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:203-096-4