2,3-Dimethyl-2,3-diphenylbutane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1889-67-4
:
Formula:H₃CC(CH₃)(C₆H₅)C(CH₃)(C₆H₅)CH₃
MDL Number:MFCD00053713
:238.37 g/mol
Boiling Pt:170…171 °C (14 mmHg)
Melting Pt:90…110 °C
Density:1,09 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:217-568-2
Specification Test Results
- Appearance:White to light yellow Crystalline powder or flakes and/or chunks
- Infrared spectrum:Conforms
- Assay:≥94 %