Order Entry
Export
Orders LinkContactUsLinkComponent
Thymol blue, pure indicator

Thymol blue, pure indicator

Thymol blue, pure indicator
Certificates
Thymol blue, pure indicator

3 Options Available Below

Product Details & Documents
Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466.6 g/mol
  • Merck Index:12,09541
  • Melting Pt:221 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-973-3

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Dark green to dark brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in ethanol) Clear
  • Specific extinction E (1%/1cm):
    ≥700 (596 nm, in 0.01 N NaOH)
  • Visual transition interval:
    from pH 1.2 (red) to pH 2.8 (yellow)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 41,80 €
    41,80 €
    1 * 5 g
       41,80 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 120,00 €
    120,00 €
    1 * 25 g
       120,00 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 363,00 €
    363,00 €
    1 * 100 g
       363,00 €