Order Entry
Export
Orders LinkContactUsLinkComponent
Phenolphthalein ACS

Phenolphthalein ACS

Phenolphthalein ACS
Certificates
Phenolphthalein ACS
Phenolphthalein ACS

2 Options Available Below

Product Details & Documents

CAS No.: 77-09-8 Molecular Formula: C20H14O4 Formula Weight: 318.33 Physical Form: Powder Density: 1.299 Melting Point: 258-262° Beilstein Reference: 284423 Merck Reference: 14,7243 MDL No.: MFCD00005913

Indicator

Synonyms:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS Number:77-09-8
  • Formula:C₂₀H₁₄O₄
  • MDL Number:MFCD00005913
  • Molecular Weight:318.33 g/mol
  • Merck Index:14,7243
  • Melting Pt:258…262 °C
  • Density:1.299
  • Storage Temperature:Ambient
  • EINECS:201-004-7
  • REACH:01-2119498295-24
Product Family Options

Product Information