4,4'-Dimethoxybenzil
Glass bottle for solids, 25 g
Catalog # MOLEM11542431
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Bis(4-methoxyphenyl)ethanedione, Anisil, p-Anisil, 1,2-Bis(4-methoxyphenyl)ethane-1,2-dione
- CAS Number:1226-42-2
- Formula:C₁₆H₁₄O₄
- MDL Number:MFCD00008405
- Molecular Weight:270.28 g/mol
- Boiling Pt:215…219 °C (3 mmHg)
- Melting Pt:128…131 °C
- Density:1.36 g/cm³ (20 °C)
- EINECS:214-960-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



