4,4'-Dimethoxybenzil 99+%
About this item
Synonyms: Bis(4-methoxyphenyl)ethanedione, Anisil, p-Anisil, 1,2-Bis(4-methoxyphenyl)ethane-1,2-dione
- CAS Number:1226-42-2
- Formula:C₁₆H₁₄O₄
- MDL Number:MFCD00008405
- Molecular Weight:270.28 g/mol
- Boiling Pt:215…219 °C (3 mmHg)
- Melting Pt:131…134 °C
- Density:1,36 g/cm³ (20 °C)
- EINECS:214-960-5
2 Options Available Below
Synonyms: Bis(4-methoxyphenyl)ethanedione, Anisil, p-Anisil, 1,2-Bis(4-methoxyphenyl)ethane-1,2-dione
- CAS Number:1226-42-2
- Formula:C₁₆H₁₄O₄
- MDL Number:MFCD00008405
- Molecular Weight:270.28 g/mol
- Boiling Pt:215…219 °C (3 mmHg)
- Melting Pt:131…134 °C
- Density:1,36 g/cm³ (20 °C)
- EINECS:214-960-5
Specifications for Product Family
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131 to 134 °C
- GC:≥99.0 %
- Residue after ignition:≤0.1 %



