Benzil
Glass bottle for solids, 4 g
Catalog # PROOICRS0222
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: 1,2-Diphenyl-1,2-ethanedione
- CAS Number:134-81-6
- Formula:(C₆H₅)C(O)C(O)(C₆H₅)
- MDL Number:MFCD00003080
- Molecular Weight:210.23 g/mol
- Merck Index:13,01078
- Boiling Pt:346 °C (1013 hPa)
- Melting Pt:95 °C
- Density:1.23 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-157-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:4 g



