2,4-Dihydroxybenzophenone, Sigma-Aldrich®
SIAL126217-100G
: Merck
:
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
:
:214.22 g/mol
:13,01107
:391 °C (1013 hPa)
:139…140 °C
:0.696 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:Ambient
:205-029-4
:131-56-6
:C₁₃H₁₀O₃
:MFCD00002277