Order Entry
Export
ContactUsLinkComponent
Bromophenol blue, Sigma-Aldrich®
Bromophenol blue, Sigma-Aldrich®
Catalog # SIALB0126-25G
Supplier:  Merck
CAS Number:  
Bromophenol blue, Sigma-Aldrich®
Catalog # SIALB0126-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :669.97 g/mol
Merck Index:13,01429
Melting Pt:273 °C
Density:1 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-086-2
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875