Bromophenol blue ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:115-39-9
:
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875
:669.97 g/mol
Merck Index:14,1444
Melting Pt:270 °C (dec.)
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-086-2