Order Entry
Export
ContactUsLinkComponent
 
Irritant
4,4'-Dibromobenzil 97%
Catalog # ACRO351780100
CAS Number:  
undefined
Irritant
4,4'-Dibromobenzil 97%
Catalog # ACRO351780100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    223 to 228 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :368.02 g/mol
Melting Pt:223…228 °C
EINECS:252-628-1
 :GHS07
CAS Number:35578-47-3
Formula:C₁₄H₈Br₂O₂
MDL Number:MFCD00000104