4-Hydroxybenzyl alcohol
Glass bottle for solids, 100 mg
Catalog # PROOMM0460.06
Supplier: LGC Standards PROMOCHEM
4-Hydroxybenzyl alcohol
Catalog # PROOMM0460.06
Supplier: LGC Standards PROMOCHEM
CAS Number: 623-05-2
Jump to:
Product Details & Documents
Synonyms: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol
- CAS Number:623-05-2
- Formula:C₇H₈O₂
- MDL Number:MFCD00004658
- Molecular Weight:124.14 g/mol
- Boiling Pt:252 °C (1013 hPa)
- Melting Pt:115…118 °C
- Storage Temperature:Refrigerator
- EINECS:210-768-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



