3,5-Dihydroxytoluene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:504-15-4
:
Formula:C₇H₈O₂
MDL Number:MFCD00002291
:124.14 g/mol
Merck Index:12,06995
Boiling Pt:290 °C (1013 hPa)
Melting Pt:109 °C
Density:1.2895 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:159 °C
Storage Temperature:Refrigerator
EINECS:207-984-2