Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hexylresorcinol
4-Hexylresorcinol
4-Hexylresorcinol
4-Hexylresorcinol
PROOMM0111.00
:
LGC Standards PROMOCHEM
:
4-Hexylresorcinol
PROOMM0111.00
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
500 mg
Seq:
2725
:
:
194.27 g/mol
:
12,04750
:
333 °C (1013 hPa)
:
64…67 °C
:
>100 °C
:
Ambient
:
205-257-4
:
GHS07
:
136-77-6
:
C₁₂H₁₈O₂
:
MFCD00002284