4-Hexylresorcinol
Catalog # MOLEM30558071
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:194.27 g/mol
Merck Index:12,04750
Boiling Pt:333 °C (1013 hPa)
Melting Pt:64…67 °C
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:205-257-4
:GHS07
CAS Number:136-77-6
Formula:C₁₂H₁₈O₂
MDL Number:MFCD00002284