
4-Hexylresorcinol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:136-77-6
:
Formula:C₁₂H₁₈O₂
MDL Number:MFCD00002284
:194.27 g/mol
Merck Index:12,04750
Boiling Pt:333 °C (1013 hPa)
Melting Pt:64…67 °C
:M-Clarity™ quality level = MQ200
Flash Pt:>100 °C
EINECS:205-257-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.5 g/l (20 °C)
- Melting Point:64 - 67 °C
- Molar Mass:194.27 g/mol
- Bulk Density:340 kg/m3
- Boiling Point:333 - 335 °C (1013 hPa)
- Flash Point:100 °C
- pH:7.7 (0.5 g/l, H2O, 20 °C)