Order Entry
Export
ContactUsLinkComponent
 
 
4-Hexylresorcinol 98%
 
undefined
 
4-Hexylresorcinol 98%

 

  • Appearance:
    White to light pink to light orange Powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    63 to 68 °C
  • HPLC:
    ≥97.5 %