2-Methylresorcinol
Glass bottle for solids, 100 g
Catalog # MOLE10864971-100G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol
- CAS Number:608-25-3
- Formula:C₇H₈O₂
- MDL Number:MFCD00002271
- Molecular Weight:124.14 g/mol
- Boiling Pt:264 °C (1013 hPa)
- Melting Pt:120…122 °C
- Flash Pt:135 °C
- Storage Temperature:Ambient
- EINECS:210-155-8
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



