2-Methylresorcinol 98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 608-25-3 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 135°(275°F) Melting Point: 118-122° Boiling Point: 264° Beilstein Reference: 2042177 UN No.: 2811 MDL No.: MFCD00002271
Synonyms: 2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol
- CAS Number:608-25-3
- Formula:C₇H₈O₂
- MDL Number:MFCD00002271
- Molecular Weight:124.14 g/mol
- Boiling Pt:264 °C
- Melting Pt:118…122 °C
- Flash Pt:135 °C (275 °F)
- Storage Temperature:Ambient
- EINECS:210-155-8
- UN:2811
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 608-25-3 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 135°(275°F) Melting Point: 118-122° Boiling Point: 264° Beilstein Reference: 2042177 UN No.: 2811 MDL No.: MFCD00002271
Synonyms: 2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol
- CAS Number:608-25-3
- Formula:C₇H₈O₂
- MDL Number:MFCD00002271
- Molecular Weight:124.14 g/mol
- Boiling Pt:264 °C
- Melting Pt:118…122 °C
- Flash Pt:135 °C (275 °F)
- Storage Temperature:Ambient
- EINECS:210-155-8
- UN:2811
- ADR:6.1,III



