Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methylresorcinol 98%

2-Methylresorcinol 98%

Toxic
2-Methylresorcinol 98%
Certificates
2-Methylresorcinol 98%

2 Options Available Below

Product Details & Documents
Synonyms:  2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol
  • CAS Number:608-25-3
  • Formula:C₇H₈O₂
  • MDL Number:MFCD00002271
  • Molecular Weight:124.14 g/mol
  • Boiling Pt:264 °C (1013 hPa)
  • Melting Pt:118…124 °C
  • Flash Pt:135 °C
  • Storage Temperature:Ambient
  • EINECS:210-155-8
  • UN:2811
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Beige to pink to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    118 to 124 °C
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 34,90 €
    34,90 €
    1 * 25 g
       34,90 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 107,00 €
    107,00 €
    1 * 100 g
       107,00 €