Order Entry
Export
Orders LinkContactUsLinkComponent
m-Cresol 99%

m-Cresol 99%

Toxic
Corrosive
m-Cresol 99%
Certificates
m-Cresol 99%

5 Options Available Below

Product Details & Documents
Synonyms:  1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene
  • CAS Number:108-39-4
  • Formula:CH₃C₆H₄OH
  • MDL Number:MFCD00002302
  • Molecular Weight:108.14 g/mol
  • Merck Index:13,02604
  • Boiling Pt:203 °C (760 mmHg)
  • Melting Pt:8…10 °C
  • Density:1.03 g/cm³
  • Flash Pt:86 °C
  • Storage Temperature:Ambient
  • EINECS:203-577-9
  • UN:2076
  • ADR:6.1,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light red Viscous liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5390 to 1.5420 (20°C, 589 nm)
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 38,10 €
    38,10 €
    1 * 100 g
       38,10 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 96,50 €
    96,50 €
    1 * 500 g
       96,50 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 172,00 €
    172,00 €
    1 * 1 kg
       172,00 €

  • Glass bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 328,00 €
    328,00 €
    1 * 2,5 kg
       328,00 €

  • Metal drum
    10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 812,00 €
    812,00 €
    1 * 10 kg
       812,00 €