Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzyl alcohol
Benzyl alcohol
Benzyl alcohol
Benzyl alcohol
EHERC10569000
:
EHRENSTORFER
:
Benzyl alcohol
EHERC10569000
:
EHRENSTORFER
:
Pack type:
Glass bottle
Pk:
1 mL
Seq:
2070
:
:
108.14 g/mol
:
13,01126
:
205 °C (1013 hPa)
:
–15.3 °C
:
1.05 g/cm³ (20 °C)
:
101 °C
:
Ambient
:
202-859-9
:
GHS07
:
100-51-6
:
C₆H₅CH₂OH
:
MFCD00004599