2-Phenylethanol, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Merck Index:12,07370
- Boiling Pt:432 °C (1013 hPa)
- Melting Pt:103…105 °C
- Density:1.02 g/cm³ (20 °C)
- Flash Pt:102 °C
- Storage Temperature:Ambient
- EINECS:200-456-2
- UN:2810
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Merck Index:12,07370
- Boiling Pt:432 °C (1013 hPa)
- Melting Pt:103…105 °C
- Density:1.02 g/cm³ (20 °C)
- Flash Pt:102 °C
- Storage Temperature:Ambient
- EINECS:200-456-2
- UN:2810
- ADR:6.1,III



