Order Entry
Export
ContactUsLinkComponent
2,6-Dimethoxytoluene
Irritant
2,6-Dimethoxytoluene
Catalog # MOLEM32379093
Supplier:  Molekula
CAS Number:  
Irritant
2,6-Dimethoxytoluene
Catalog # MOLEM32379093
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.19 g/mol
Boiling Pt:52…56 °C (0.5 mmHg)
Boiling Pt:52…56 °C (0,5 mmHg)
Melting Pt:39 °C
EINECS:227-131-8
 :GHS07
CAS Number:5673-07-4
Formula:C₉H₁₂O₂
MDL Number:MFCD00008374