


2,4-Dinitrotoluene
Catalog # EHERC12786200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:182.14 g/mol
Boiling Pt:300 °C (1013 hPa)
Melting Pt:67…70 °C
Density:1.379 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-450-0
UN:3454
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:121-14-2
Formula:C₇H₆N₂O₄
MDL Number:MFCD00007172