


2,6-Diaminotoluene
Catalog # EHERC12197800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:122.17 g/mol
Boiling Pt:280…282 °C (1013 hPa)
Melting Pt:113 °C
EINECS:212-513-9
UN:2811
ADR:6.1,III
:GHS08
:GHS07
:GHS09
CAS Number:823-40-5
Formula:C₇H₁₀N₂
MDL Number:MFCD00007800