Order Entry
Export
ContactUsLinkComponent
3,4-Diaminotoluene
3,4-Diaminotoluene
Catalog # EHERC12197900
Supplier:  EHRENSTORFER
CAS Number:  
3,4-Diaminotoluene
Catalog # EHERC12197900
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :122.17 g/mol
Boiling Pt:265 °C (1013 hPa)
Melting Pt:86…89 °C
Storage Temperature:Ambient
EINECS:207-826-2
CAS Number:496-72-0
Formula:C₇H₁₀N₂
MDL Number:MFCD00007728