


2,3-Diaminotoluene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2687-25-4
:
Formula:C₇H₁₀N₂
MDL Number:MFCD00011589
:122.17 g/mol
Boiling Pt:255 °C (1013 hPa)
Melting Pt:58…63 °C
Density:1,3 g/cm³ (20 °C)
EINECS:220-248-5
UN:2811
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Brown to dark brown Powder or crystals
- Infrared spectrum:Conforms
- Melting point:58 to 63 °C (Dec)
- HPLC:≥96.0 %
- Titration with HClO4:≥96.0 %