Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methoxybenzylamine

2-Methoxybenzylamine

Corrosive
Supplier:   CHEMPUR
2-Methoxybenzylamine
Supplier:  CHEMPUR
Certificates
2-Methoxybenzylamine

2 Options Available Below

Product Details & Documents
Synonyms:  (2-Methoxyphenyl)methanamine, 2-(Aminomethyl)anisole, o-Anisylamine
  • CAS Number:6850-57-3
  • Formula:C₈H₁₁NO
  • MDL Number:MFCD00008110
  • Molecular Weight:137.18 g/mol
  • Boiling Pt:227 °C (1013 hPa)
  • Density:1.05 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:229-941-7
  • UN:2735
  • ADR:8,III
  •  :Density: 1,05 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

This product has been blocked by your organization. Please contact your purchasing department for more information