

N,N-Dimethyl-p-toluidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-97-8
:
Formula:C₉H₁₃N
MDL Number:MFCD00008316
:135.21 g/mol
Boiling Pt:211 °C (1013 hPa)
Melting Pt:–15 °C
Density:0.9 g/cm³ (20 °C)
Flash Pt:76 °C
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:202-805-4
:GHS06
:GHS08
Storage Temperature:Ambient