

N,N-Dimethyl-p-toluidine, Sigma-Aldrich®
Catalog # SIAL41700-1L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:135.21 g/mol
Boiling Pt:211 °C (1013 hPa)
Melting Pt:–15 °C
Density:0.9 g/cm³ (20 °C)
Flash Pt:76 °C
Storage Temperature:Ambient
EINECS:202-805-4
UN:1708
ADR:6.1,II
:M-Clarity™ quality level = MQ100
:GHS06
:GHS08
CAS Number:99-97-8
Formula:C₉H₁₃N
MDL Number:MFCD00008316