α,α,α-Trifluoro-o-toluidine
Catalog # MOLE59569851-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Boiling Pt:175 °C (1013 hPa)
Melting Pt:–33 °C
Density:1.282 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,III
:161.13 g/mol
EINECS:201-806-7
UN:2942
CAS Number:88-17-5
Formula:C₇H₆F₃N
MDL Number:MFCD00007718