Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,α,α-Trifluoro-o-toluidine
α,α,α-Trifluoro-o-toluidine ≥98%
α,α,α-Trifluoro-o-toluidine ≥98%
:
Thermo Fisher Scientific
α,α,α-Trifluoro-o-toluidine ≥98%
:
Thermo Fisher Scientific
:
88-17-5
:
:
C₇H₆F₃N
:
MFCD00007718
:
67…68 °C (15 mmHg)
:
–35…–33 °C
:
1.295
:
65 °C (149 °F)
:
Ambient
:
201-806-7
:
6.1,III
:
161.13 g/mol
:
2942
:
GHS05
:
GHS08
:
GHS07
msds_ALFAA15072
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years