(2,4-Difluorophenyl)acetonitrile, Sigma-Aldrich®
Catalog # SIAL264490-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:153.13 g/mol
Boiling Pt:90 °C (0.3 mmHg)
Density:1.2503 g/cm³ (15 °C)
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:211-514-1
UN:3276
CAS Number:656-35-9
Formula:C₈H₅F₂N
MDL Number:MFCD00009971