


3-Chloro-p-cresol ≥97%
Catalog # H27385.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:615-62-3
:
Formula:C₇H₇ClO
MDL Number:MFCD00060319
:142.58 g/mol
Boiling Pt:228 °C
Melting Pt:51…53 °C
EINECS:210-439-1
UN:3437
ADR:6.1,II
:GHS06
:GHS05
:GHS07
Storage Temperature:Ambient