Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-p-cresol
2-Chloro-p-cresol
2-Chloro-p-cresol
EHERC11439200
:
EHRENSTORFER
:
2-Chloro-p-cresol
EHERC11439200
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
:
:
198…203 °C (1013 hPa)
:
76 °C
:
1.211 g/cm³ (25 °C)
:
6.1,II
:
142.58 g/mol
:
229-656-8
:
2669
:
GHS07
:
6640-27-3
:
C₇H₇ClO
:
MFCD00070639