Order Entry
Export
Orders LinkContactUsLinkComponent
2-Chloro-p-toluidine

2-Chloro-p-toluidine

Glass bottle for solids, 25 g
Catalog # MOLEM17655382
Irritant
Supplier:   Molekula
2-Chloro-p-toluidine
Catalog # MOLEM17655382
Supplier:  Molekula
CAS Number:  615-65-6
2-Chloro-p-toluidine

Glass bottle for solids, 25 g

CAS Number:  615-65-6
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Chloro-4-methylaniline, 4-Amino-3-chlorotoluene
  • CAS Number:615-65-6
  • Formula:C₇H₈ClN
  • MDL Number:MFCD00007666
  • Molecular Weight:141.6 g/mol
  • Boiling Pt:224 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:1.151 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:210-440-7
  • UN:3429
  • ADR:6.1,III
  •  :Density: 1.151 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications