Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dichlorobenzylamine
3,4-Dichlorobenzylamine 96%
3,4-Dichlorobenzylamine 96%
:
Thermo Fisher Scientific
3,4-Dichlorobenzylamine 96%
:
Thermo Fisher Scientific
:
102-49-8
:
:
C₇H₇Cl₂N
:
MFCD00008114
:
176.04 g/mol
:
80…82 °C (0.6 mmHg)
:
1.322
:
>110 °C (230 °F)
:
Ambient
:
203-035-1
:
2735
:
8,III
:
GHS05
msds_ALFAL04899
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years