Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichlorobenzylamine
2,6-Dichlorobenzylamine ≥97%
2,6-Dichlorobenzylamine ≥97%
:
Thermo Fisher Scientific
2,6-Dichlorobenzylamine ≥97%
:
Thermo Fisher Scientific
:
6575-27-5
:
:
C₇H₇Cl₂N
:
MFCD00047928
:
176.04 g/mol
:
Ambient
:
2735
:
8,III
:
GHS05
msds_ALFAH29274
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years