

2,6-Dichloro-4-(trifluoromethyl)aniline
Catalog # MAYBSEW01282.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:230.02 g/mol
Boiling Pt:60…62 °C (1 mmHg)
Melting Pt:32…35 °C
Density:1.532 g/cm³ (20 °C)
EINECS:416-430-0
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:24279-39-8
Formula:C₇H₄Cl₂F₃N
MDL Number:MFCD00052918