
2,4-Dichlorobenzotrifluoride, Sigma-Aldrich®
Catalog # SIAL152250-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:215 g/mol
Boiling Pt:193 °C (1013 hPa)
Melting Pt:–1 °C
Density:1.4929 g/cm³ (20 °C)
EINECS:206-278-1
UN:3265
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:320-60-5
Formula:C₇H₃Cl₂F₃
MDL Number:MFCD00000582