Order Entry
Export
ContactUsLinkComponent
(4-Chlorophenyl)acetonitrile, Sigma-Aldrich®
(4-Chlorophenyl)acetonitrile, Sigma-Aldrich®
Catalog # SIALC28006-100G
Supplier:  Merck
CAS Number:  
(4-Chlorophenyl)acetonitrile, Sigma-Aldrich®
Catalog # SIALC28006-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :151.6 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:30.5 °C
Density:1.1968 g/cm³ (20 °C)
Flash Pt:133 °C
Storage Temperature:Ambient
EINECS:205-418-9
UN:3439
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
CAS Number:140-53-4
Formula:C₈H₆ClN
MDL Number:MFCD00001918