(4-Chlorophenyl)acetonitrile
Catalog # MOLE16706246-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:151.6 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:30.5 °C
Density:1.1968 g/cm³ (20 °C)
Flash Pt:133 °C
Storage Temperature:Ambient
EINECS:205-418-9
UN:3439
ADR:6.1,II
CAS Number:140-53-4
Formula:C₈H₆ClN
MDL Number:MFCD00001918