Order Entry
Export
ContactUsLinkComponent
(4-Chlorophenyl)acetonitrile 98+%
Toxic
(4-Chlorophenyl)acetonitrile 98+%
Catalog # ACRO108810050
CAS Number:  
undefined
Toxic
(4-Chlorophenyl)acetonitrile 98+%
Catalog # ACRO108810050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5415 to 1.5435 (20°C, 589 nm)
  • Water:
    ≤0.3 % (K.F.)
  • GC:
    ≥98.0 %

Specification Test Results

About this item

 :
 :151.6 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:29…30 °C
Density:1,1968 g/cm³ (20 °C)
Flash Pt:133 °C
Storage Temperature:Ambient
EINECS:205-418-9
UN:3439
ADR:6.1,II
 :GHS06
CAS Number:140-53-4
Formula:C₈H₆ClN
MDL Number:MFCD00001918